Assay Details
| Gene Symbol: CYP24A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214075.2 | NP_999240.2 | 6 - 7 | 1218 | 63 | |
| GenBank mRNA | AF245504.2 | - | 6 - 7 | 1236 | 63 | |
| AK393511.1 | - | 6 - 7 | 1248 | 63 | ||
| AK393556.1 | - | 5 - 6 | 567 | 63 | ||
More Information
Set Membership: |
|

CYP24A1
Genomic Map