Assay Details
| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214326.2 | NP_999491.1 | 2 - 3 | 165 | 56 | |
| GenBank mRNA | AB055884.1 | - | 2 - 3 | 186 | 56 | |
| AK240171.1 | - | 2 - 3 | 183 | 56 | ||
| AK391832.1 | - | 2 - 3 | 182 | 56 | ||
| AY550044.1 | - | 1 - 2 | 141 | 56 | ||
| DQ629168.1 | - | 2 - 3 | 174 | 56 | ||
More Information
Set Membership: |
|

RPL35
Genomic Map