Assay Details
| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214326.2 | NP_999491.1 | 3 - 4 | 246 | 67 | |
| GenBank mRNA | AB055884.1 | - | 3 - 4 | 267 | 67 | |
| AK240171.1 | - | 3 - 4 | 264 | 67 | ||
| AK391832.1 | - | 3 - 4 | 263 | 67 | ||
| AY550044.1 | - | 2 - 3 | 222 | 67 | ||
More Information
Set Membership: |
|

RPL35
Genomic Map