Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001634.1 | NP_001001634.1 | 1 - 2 | 155 | 86 | |
GenBank mRNA | AK239565.1 | - | 1 | 179 | 86 | |
AK344835.1 | - | 2 - 3 | 180 | 86 | ||
AK352159.1 | - | 1154 | 86 | |||
AY550073.1 | - | 1 - 2 | 155 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
