Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001636.1 | NP_001001636.1 | 3 - 4 | 297 | 94 | |
| XM_021068582.1 | XP_020924241.1 | 384 | 94 | |||
| GenBank mRNA | AK392319.1 | - | 3 - 4 | 324 | 94 | |
| AK400064.1 | - | 3 - 4 | 364 | 94 | ||
| AY550039.1 | - | 3 - 4 | 297 | 94 | ||
| AY610304.1 | - | 2 - 3 | 279 | 94 | ||
More Information
Set Membership: |
|

RPL32
Genomic Map