Assay Details
Gene Symbol: APOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002801.1 | NP_001002801.1 | 1 - 2 | 55 | 102 | |
XM_005667372.2 | XP_005667429.1 | 158 | 102 | |||
XM_021102247.1 | XP_020957906.1 | 272 | 102 | |||
XM_021102248.1 | XP_020957907.1 | 210 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
