Assay Details
Gene Symbol: NAA80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003663.1 | NP_001003663.1 | 1 | 545 | 71 |
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK343314.1 | - | 2 | 661 | 71 | |
AK346555.1 | - | 2 | 661 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
