Assay Details
| Gene Symbol: ANGPTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003926.2 | NP_001003926.1 | 2 - 3 | 628 | 104 | |
| GenBank mRNA | AY523565.2 | - | 2 - 3 | 630 | 104 | |
More Information
Set Membership: |
|

ANGPTL3
Genomic Map