Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004046.2 | NP_001004046.1 | 2 - 3 | 242 | 73 | |
| GenBank mRNA | X77640.1 | - | 247 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004046.2 | NP_001004046.1 | 2 - 3 | 242 | 73 | |
| GenBank mRNA | X77640.1 | - | 247 | 73 | ||
Set Membership: |
|