Assay Details
| Gene Symbol: NFKBIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005150.1 | NP_001005150.1 | 5 - 6 | 895 | 65 | |
| GenBank mRNA | AK233732.1 | - | 4 - 5 | 659 | 65 | |
| AK239662.1 | - | 6 - 7 | 1155 | 65 | ||
| AK389460.1 | - | 4 - 5 | 1289 | 65 | ||
| AK395569.1 | - | 5 - 6 | 1271 | 65 | ||
| AK395997.1 | - | 4 - 5 | 1357 | 65 | ||
| AK397107.1 | - | 5 - 6 | 1041 | 65 | ||
| AK399004.1 | - | 5 - 6 | 1020 | 65 | ||
| AK399095.1 | - | 4 - 5 | 900 | 65 | ||
| AK399188.1 | - | 5 - 6 | 1006 | 65 | ||
| AY609599.1 | - | 5 - 6 | 992 | 65 | ||
| FN421467.1 | - | 5 - 6 | 832 | 65 | ||
More Information
Set Membership: |
|

NFKBIA
Genomic Map