Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009580.1 | NP_001009580.1 | 2 - 3 | 231 | 70 | |
| XM_005671243.3 | XP_005671300.1 | 278 | 70 | |||
| XM_005671244.3 | XP_005671301.1 | 300 | 70 | |||
| XM_021072178.1 | XP_020927837.1 | 278 | 70 | |||
| GenBank mRNA | AJ577083.1 | - | 2 - 3 | 231 | 70 | |
| AK233811.1 | - | 2 - 3 | 247 | 70 | ||
| AK235057.1 | - | 2 - 3 | 263 | 70 | ||
| AK240146.1 | - | 2 - 3 | 254 | 70 | ||
| AK394475.1 | - | 2 - 3 | 250 | 70 | ||
| AK395393.1 | - | 2 - 3 | 279 | 70 | ||
| AY312066.1 | - | 2 - 3 | 231 | 70 | ||
| AY610254.1 | - | 2 - 3 | 239 | 70 | ||
| DQ124105.1 | - | 2 - 3 | 191 | 70 | ||
More Information
Set Membership: |
|

CXCL12
Genomic Map