Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011508.2 | NP_001011508.1 | 6 - 7 | 1637 | 63 | |
| XM_021077321.1 | XP_020932980.1 | 1612 | 63 | |||
| XM_021077322.1 | XP_020932981.1 | 1407 | 63 | |||
| XM_021077323.1 | XP_020932982.1 | 1606 | 63 | |||
| XM_021077324.1 | XP_020932983.1 | 1275 | 63 | |||
| GenBank mRNA | AJ544724.1 | - | 6 - 7 | 1637 | 63 | |
| KJ820675.1 | - | 6 - 7 | 1637 | 63 | ||
More Information
Set Membership: |
|

SIRPA
Genomic Map