Assay Details
| Gene Symbol: AURKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025225.1 | NP_001020396.1 | 2 - 3 | 318 | 77 | |
| XM_005673034.3 | XP_005673091.1 | 721 | 77 | |||
| XM_005673035.3 | XP_005673092.1 | 571 | 77 | |||
| XM_005673036.3 | XP_005673093.1 | 571 | 77 | |||
| XM_021077325.1 | XP_020932984.1 | 445 | 77 | |||
| XM_021077326.1 | XP_020932985.1 | 431 | 77 | |||
| GenBank mRNA | AB196773.1 | - | 2 - 3 | 318 | 77 | |
| EF486358.1 | - | 2 - 3 | 333 | 77 | ||
More Information
Set Membership: |
|

AURKA
Genomic Map