Assay Details
| Gene Symbol: FABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025229.1 | NP_001020400.1 | 1 - 2 | 149 | 73 | |
| GenBank mRNA | AK391779.1 | - | 1 - 2 | 167 | 73 | |
| AY610347.1 | - | 1 - 2 | 149 | 73 | ||
| DQ000160.1 | - | 1 - 2 | 149 | 73 | ||
More Information
Set Membership: |
|

FABP7
Genomic Map