Assay Details
Gene Symbol: PTGES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038631.1 | NP_001033720.1 | 1 - 2 | 217 | 70 | |
GenBank mRNA | AK231896.1 | - | 1 - 2 | 199 | 70 | |
AK348546.1 | - | 1 - 2 | 165 | 70 | ||
AK392435.1 | - | 1 - 2 | 200 | 70 | ||
AK399226.1 | - | 1 - 2 | 197 | 70 | ||
AY857634.1 | - | 1 - 2 | 217 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
