Assay Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039746.2 | NP_001034835.1 | 3 - 4 | 433 | 72 | |
GenBank mRNA | AB529462.1 | - | 3 - 4 | 351 | 72 | |
AK231381.1 | - | 3 - 4 | 434 | 72 | ||
AK346851.1 | - | 591 | 72 | |||
AY841270.1 | - | 3 - 4 | 436 | 72 | ||
DQ372083.1 | - | 3 - 4 | 434 | 72 | ||
DQ523617.1 | - | 3 - 4 | 318 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
