Assay Details
| Gene Symbol: SLA-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213768.1 | NP_998933.1 | 2 - 3 | 339 | 70 | |
| XM_005665788.3 | XP_005665845.2 | 514 | 70 | |||
| XM_013977626.2 | XP_013833080.2 | 484 | 70 | |||
| XM_021098282.1 | XP_020953941.1 | 797 | 70 | |||
| GenBank mRNA | AY463541.1 | - | 2 - 3 | 339 | 70 | |
| GU322918.1 | - | 2 - 3 | 70 | |||
| HQ224544.1 | - | 2 - 3 | 339 | 70 | ||
| HQ224545.1 | - | 2 - 3 | 339 | 70 | ||
| HQ224546.1 | - | 2 - 3 | 317 | 70 | ||
| HQ224547.1 | - | 2 - 3 | 317 | 70 | ||
More Information
Set Membership: |
|

SLA-7
Genomic Map