Assay Details
| Gene Symbol: SLA-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213768.1 | NP_998933.1 | 6 - 7 | 1034 | 80 | |
| XM_005665788.3 | XP_005665845.2 | 1209 | 80 | |||
| XM_013977626.2 | XP_013833080.2 | 1179 | 80 | |||
| XM_021098282.1 | XP_020953941.1 | 1492 | 80 | |||
| GenBank mRNA | AY463541.1 | - | 6 - 7 | 1034 | 80 | |
| GU322919.1 | - | 3 - 4 | 219 | 80 | ||
More Information
Set Membership: |
|

SLA-7
Genomic Map