Assay Details
Gene Symbol: CXCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213773.1 | NP_998938.1 | 2 | 950 | 94 | |
GenBank mRNA | AB116560.1 | - | 2 | 950 | 94 | |
AK238891.1 | - | 2 | 1038 | 94 | ||
AK238923.1 | - | 2 | 1152 | 94 | ||
AK351860.1 | - | 2 | 1038 | 94 | ||
AK389753.1 | - | 2 | 1040 | 94 | ||
AK399441.1 | - | 2 | 1040 | 94 | ||
DQ124104.1 | - | 2 | 950 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
