Assay Details
| Gene Symbol: GHRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213807.2 | NP_998972.1 | 2 - 3 | 257 | 64 | |
| XM_013981924.2 | XP_013837378.1 | 2860 | 64 | |||
| GenBank mRNA | AB035703.1 | - | 2 - 3 | 243 | 64 | |
| AB035704.1 | - | 2 - 3 | 241 | 64 | ||
| AF308930.1 | - | 2 - 3 | 258 | 64 | ||
| AY609454.1 | - | 2 - 3 | 245 | 64 | ||
| DQ355969.1 | - | 2 - 3 | 258 | 64 | ||
More Information
Set Membership: |
|

GHRL
Genomic Map