Assay Details
Gene Symbol: PPAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213809.1 | NP_998974.1 | 7 - 8 | 921 | 77 | |
GenBank mRNA | AF315377.1 | - | 7 - 8 | 921 | 77 | |
AY188554.1 | - | 6 - 7 | 780 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
