Assay Details
Gene Symbol: SAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213814.2 | NP_998979.1 | 5 - 6 | 582 | 121 | |
GenBank mRNA | AJ249974.1 | - | 5 - 6 | 582 | 121 | |
AK350053.1 | - | 5 - 6 | 616 | 121 | ||
AK399206.1 | - | 5 - 6 | 608 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
