Assay Details
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213852.1 | NP_999017.1 | 2 - 3 | 149 | 79 | |
GenBank mRNA | AK230778.1 | - | 3 - 4 | 293 | 79 | |
AK233739.1 | - | 3 - 4 | 678 | 79 | ||
S80724.1 | - | 2 - 3 | 149 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
