Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213911.1 | NP_999076.1 | 3 | 548 | 91 | |
| GenBank mRNA | AK233188.1 | - | 3 | 381 | 91 | |
| AK235537.1 | - | 2 | 166 | 91 | ||
| X99714.1 | - | 548 | 91 | |||
More Information
Set Membership: |
|

PGRMC1
Genomic Map