Assay Details
| Gene Symbol: C4BPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213942.1 | NP_999107.1 | 3 | 349 | 111 | |
| GenBank mRNA | AF143229.1 | - | 3 | 111 | ||
| AF143230.1 | - | 3 | 295 | 111 | ||
| AK238136.1 | - | 3 | 552 | 111 | ||
| AK346515.1 | - | 3 | 383 | 111 | ||
| L06820.1 | - | 3 | 349 | 111 | ||
More Information
Set Membership: |
|

C4BPA
Genomic Map