Assay Details
| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213956.1 | NP_999121.1 | 8 | 1749 | 58 | |
| XM_021066204.1 | XP_020921863.1 | 2015 | 58 | |||
| XM_021066205.1 | XP_020921864.1 | 1729 | 58 | |||
| XM_021066206.1 | XP_020921865.1 | 1967 | 58 | |||
| GenBank mRNA | X69675.1 | - | 1749 | 58 | ||
More Information
Set Membership: |
|

HNF1B
Genomic Map