Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213958.1 | NP_999123.1 | 2 | 2853 | 63 | |
| XM_005669564.3 | XP_005669621.2 | 3844 | 63 | |||
| GenBank mRNA | AB071394.1 | - | 2 | 2853 | 63 | |
| AK349013.1 | - | 2 | 2864 | 63 | ||
| AY373817.1 | - | 1 | 516 | 63 | ||
| AY859728.1 | - | 2 | 2875 | 63 | ||
| GU138029.1 | - | 2 | 2875 | 63 | ||
| JX232221.1 | - | 1 | 849 | 63 | ||
| JX307113.1 | - | 1 | 2799 | 63 | ||
| KC860785.1 | - | 1 | 2799 | 63 | ||
More Information
Set Membership: |
|

TLR9
Genomic Map