Assay Details
| Gene Symbol: SELENOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213977.1 | NP_999142.1 | 5 | 250 | 107 | |
| GenBank mRNA | AF380118.1 | - | 5 | 250 | 107 | |
| AK347199.1 | - | 5 | 267 | 107 | ||
| AK394798.1 | - | 5 | 299 | 107 | ||
More Information
Set Membership: |
|

SELENOW
Genomic Map