Assay Details
| Gene Symbol: KCNN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214018.2 | NP_999183.1 | 7 - 8 | 1249 | 64 | |
| GenBank mRNA | AY062036.1 | - | 7 - 8 | 1251 | 64 | |
More Information
Set Membership: |
|

KCNN4
Genomic Map