Assay Details
| Gene Symbol: TBXAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214046.1 | NP_999211.1 | 9 - 10 | 1152 | 65 | |
| XM_021078508.1 | XP_020934167.1 | 1455 | 65 | |||
| XM_021078509.1 | XP_020934168.1 | 1644 | 65 | |||
| GenBank mRNA | L13128.1 | - | 9 - 10 | 1152 | 65 | |
More Information
Set Membership: |
|

TBXAS1
Genomic Map