Assay Details
| Gene Symbol: IL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214055.1 | NP_999220.1 | 4 - 5 | 368 | 96 | |
| XM_021085847.1 | XP_020941506.1 | 669 | 96 | |||
| GenBank mRNA | AK344440.1 | - | 4 - 5 | 386 | 96 | |
| M86725.1 | - | 4 - 5 | 368 | 96 | ||
More Information
Set Membership: |
|

IL1B
Genomic Map