Assay Details
| Gene Symbol: DAO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214066.2 | NP_999231.1 | 10 - 11 | 1123 | 61 | |
| GenBank mRNA | AK390956.1 | - | 10 - 11 | 1105 | 61 | |
| AK391151.1 | - | 10 - 11 | 996 | 61 | ||
| M16972.1 | - | 10 - 11 | 1123 | 61 | ||
| M18448.1 | - | 5 - 6 | 456 | 61 | ||
More Information
Set Membership: |
|

DAO
Genomic Map