Assay Details
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214091.2 | NP_999256.1 | 3 - 4 | 397 | 67 | |
GenBank mRNA | AF484233.1 | - | 3 - 4 | 398 | 67 | |
AF484234.1 | - | 2 - 3 | 275 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
