Assay Details
Gene Symbol: PPARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130241.3 | NP_001123713.1 | 4 - 5 | 658 | 72 | |
NM_214152.3 | NP_999317.2 | 5 - 6 | 742 | 72 | ||
XM_005665910.3 | XP_005665967.1 | 823 | 72 | |||
XM_013977808.2 | XP_013833262.1 | 1447 | 72 | |||
GenBank mRNA | AY188501.1 | - | 5 - 6 | 708 | 72 | |
GU565977.1 | - | 3 - 4 | 528 | 72 | ||
KP757025.1 | - | 3 - 4 | 427 | 72 | ||
KP757026.1 | - | 3 - 4 | 427 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
