Assay Details
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214216.1 | NP_999381.1 | 3 - 4 | 298 | 85 | |
XM_021088713.1 | XP_020944372.1 | 413 | 85 | |||
XM_021088714.1 | XP_020944373.1 | 465 | 85 | |||
XM_021088715.1 | XP_020944374.1 | 3443 | 85 | |||
GenBank mRNA | AF056489.1 | - | 3 - 4 | 298 | 85 | |
AK344681.1 | - | 4 - 5 | 412 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
