Assay Details
| Gene Symbol: PRKAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214266.1 | NP_999431.1 | 7 - 8 | 1336 | 131 | |
| XM_021093564.1 | XP_020949223.1 | 1149 | 131 | |||
| GenBank mRNA | AK235206.1 | - | 7 - 8 | 1319 | 131 | |
| AK348146.1 | - | 7 - 8 | 1369 | 131 | ||
| AK390267.1 | - | 7 - 8 | 1368 | 132 | ||
| AY159788.1 | - | 7 - 8 | 1336 | 131 | ||
More Information
Set Membership: |
|

PRKAA2
Genomic Map