Assay Details
| Gene Symbol: VCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214280.1 | NP_999445.1 | 3 - 4 | 478 | 73 | |
| XM_021083237.1 | XP_020938896.1 | 683 | 73 | |||
| XM_021083238.1 | XP_020938897.1 | 656 | 73 | |||
| GenBank mRNA | AK346150.1 | - | 3 - 4 | 563 | 73 | |
| AK348383.1 | - | 3 - 4 | 605 | 73 | ||
| AY609417.1 | - | 3 - 4 | 483 | 73 | ||
| M30143.1 | - | 3 - 4 | 478 | 73 | ||
More Information
Set Membership: |
|

VCP
Genomic Map