Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214364.1 | NP_999529.1 | 5 - 6 | 544 | 62 | |
| XM_021077307.1 | XP_020932966.1 | 698 | 62 | |||
| XM_021077308.1 | XP_020932967.1 | 767 | 62 | |||
| GenBank mRNA | AF268463.1 | - | 5 - 6 | 544 | 62 | |
| AK344428.1 | - | 6 - 7 | 601 | 62 | ||
| JX280457.1 | - | 5 - 6 | 511 | 62 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map