Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214433.1 | NP_999598.1 | 2 - 3 | 294 | 72 | |
XM_005665807.3 | XP_005665864.1 | 449 | 72 | |||
XM_021098323.1 | XP_020953982.1 | 452 | 72 | |||
XM_021098325.1 | XP_020953984.1 | 448 | 72 | |||
GenBank mRNA | AK343239.1 | - | 2 - 3 | 398 | 72 | |
AK343420.1 | - | 2 - 3 | 340 | 72 | ||
AK389200.1 | - | 1 - 2 | 106 | 72 | ||
AK389399.1 | - | 2 - 3 | 306 | 72 | ||
AK389523.1 | - | 2 - 3 | 310 | 72 | ||
AK398798.1 | - | 2 - 3 | 304 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
