Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214433.1 | NP_999598.1 | 3 - 4 | 449 | 119 | |
XM_005665806.3 | XP_005665863.1 | 717 | 119 | |||
XM_005665807.3 | XP_005665864.1 | 604 | 119 | |||
XM_021098321.1 | XP_020953980.1 | 723 | 119 | |||
XM_021098322.1 | XP_020953981.1 | 720 | 119 | |||
XM_021098323.1 | XP_020953982.1 | 607 | 119 | |||
XM_021098325.1 | XP_020953984.1 | 603 | 119 | |||
GenBank mRNA | AK343239.1 | - | 3 - 4 | 553 | 119 | |
AK343364.1 | - | 3 - 4 | 490 | 119 | ||
AK343420.1 | - | 3 - 4 | 495 | 119 | ||
AK343514.1 | - | 2 - 3 | 575 | 119 | ||
AK389200.1 | - | 2 - 3 | 261 | 119 | ||
AK389399.1 | - | 3 - 4 | 461 | 119 | ||
AK389523.1 | - | 3 - 4 | 465 | 119 | ||
AK398798.1 | - | 3 - 4 | 459 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
