Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214433.1 | NP_999598.1 | 4 - 5 | 542 | 69 | |
| XM_005665806.3 | XP_005665863.1 | 810 | 69 | |||
| XM_005665807.3 | XP_005665864.1 | 697 | 69 | |||
| XM_021098321.1 | XP_020953980.1 | 816 | 69 | |||
| XM_021098322.1 | XP_020953981.1 | 813 | 69 | |||
| XM_021098323.1 | XP_020953982.1 | 700 | 69 | |||
| XM_021098325.1 | XP_020953984.1 | 696 | 69 | |||
| GenBank mRNA | AK343239.1 | - | 4 - 5 | 646 | 69 | |
| AK343364.1 | - | 4 - 5 | 583 | 69 | ||
| AK343420.1 | - | 4 - 5 | 588 | 69 | ||
| AK343514.1 | - | 3 - 4 | 668 | 69 | ||
| AK389200.1 | - | 3 - 4 | 354 | 69 | ||
| AK389399.1 | - | 4 - 5 | 554 | 69 | ||
| AK389523.1 | - | 4 - 5 | 558 | 69 | ||
More Information
Set Membership: |
|

CYP21A2
Genomic Map