Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214433.1 | NP_999598.1 | 5 - 6 | 644 | 94 | |
| XM_005665806.3 | XP_005665863.1 | 912 | 94 | |||
| XM_021098322.1 | XP_020953981.1 | 915 | 94 | |||
| XM_021098325.1 | XP_020953984.1 | 798 | 94 | |||
| GenBank mRNA | AK343239.1 | - | 5 - 6 | 748 | 94 | |
| AK343364.1 | - | 5 - 6 | 685 | 94 | ||
| AK343388.1 | - | 1 - 2 | 274 | 94 | ||
| AK343420.1 | - | 5 - 6 | 690 | 94 | ||
| AK343514.1 | - | 4 - 5 | 770 | 94 | ||
| AK389200.1 | - | 4 - 5 | 456 | 94 | ||
| AK389399.1 | - | 5 - 6 | 656 | 94 | ||
| AK389523.1 | - | 5 - 6 | 660 | 94 | ||
More Information
Set Membership: |
|

CYP21A2
Genomic Map