Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214433.1 | NP_999598.1 | 6 - 7 | 730 | 115 | |
| GenBank mRNA | AK343239.1 | - | 6 - 7 | 834 | 115 | |
| AK343420.1 | - | 6 - 7 | 776 | 115 | ||
| AK389200.1 | - | 5 - 6 | 542 | 115 | ||
| AK389308.1 | - | 2 - 3 | 111 | 115 | ||
| AK389429.1 | - | 2 - 3 | 111 | 115 | ||
| AK398753.1 | - | 2 - 3 | 111 | 115 | ||
| AK398798.1 | - | 5 - 6 | 842 | 115 | ||
More Information
Set Membership: |
|

CYP21A2
Genomic Map