Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214433.1 | NP_999598.1 | 8 - 9 | 1114 | 81 | |
GenBank mRNA | AK343239.1 | - | 8 - 9 | 1218 | 81 | |
AK343420.1 | - | 8 - 9 | 1160 | 81 | ||
AK389200.1 | - | 7 - 8 | 926 | 81 | ||
AK389308.1 | - | 4 - 5 | 495 | 81 | ||
AK389429.1 | - | 4 - 5 | 495 | 81 | ||
AK398798.1 | - | 7 - 8 | 1226 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
