Assay Details
| Gene Symbol: HYAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214439.1 | NP_999604.1 | 2 - 3 | 1002 | 123 | |
| XM_005669570.3 | XP_005669627.1 | 1541 | 123 | |||
| XM_005669571.3 | XP_005669628.1 | 1087 | 123 | |||
| GenBank mRNA | AK349560.1 | - | 3 - 4 | 1062 | 123 | |
| AY497545.1 | - | 2 - 3 | 1002 | 123 | ||
More Information
Set Membership: |
|

HYAL3
Genomic Map