Assay Details
| Gene Symbol: HYAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214441.1 | NP_999606.1 | 2 - 3 | 1538 | 65 | |
| GenBank mRNA | AK231433.1 | - | 3 - 4 | 1125 | 65 | |
| AY497543.1 | - | 2 - 3 | 1538 | 65 | ||
| EU925133.1 | - | 2 - 3 | 1048 | 65 | ||
More Information
Set Membership: |
|

HYAL1
Genomic Map