Assay Details
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044544.1 | NP_001038009.1 | 4 | 1349 | 143 | |
| GenBank mRNA | AK232738.1 | - | 4 | 1326 | 143 | |
| AK232778.1 | - | 4 | 1326 | 143 | ||
| AK233575.1 | - | 4 | 1326 | 143 | ||
| AK234324.1 | - | 4 | 1326 | 143 | ||
| AK235050.1 | - | 4 | 1327 | 143 | ||
| AK235303.1 | - | 4 | 135 | |||
| AK237110.1 | - | 4 | 1326 | 143 | ||
| AK237177.1 | - | 5 | 1161 | 143 | ||
| AK238447.1 | - | 4 | 1326 | 143 | ||
| AK238448.1 | - | 4 | 1326 | 143 | ||
| AK238573.1 | - | 4 | 1326 | 143 | ||
| AK238575.1 | - | 4 | 1326 | 143 | ||
| AK238602.1 | - | 4 | 1326 | 143 | ||
| AK239609.1 | - | 4 | 1326 | 143 | ||
| AK239809.1 | - | 4 | 1262 | 143 | ||
| AK239824.1 | - | 3 | 912 | 143 | ||
| AK348545.1 | - | 5 | 1264 | 143 | ||
| AK351304.1 | - | 4 | 1329 | 143 | ||
| AK392302.1 | - | 4 | 1327 | 143 | ||
| AK396000.1 | - | 4 | 1326 | 143 | ||
| AK398002.1 | - | 4 | 1326 | 143 | ||
| AK398119.1 | - | 4 | 143 | |||
| AK398124.1 | - | 4 | 1326 | 143 | ||
| AK398171.1 | - | 4 | 1326 | 143 | ||
| AK398193.1 | - | 4 | 1326 | 143 | ||
| AK398196.1 | - | 4 | 1326 | 143 | ||
| AK398227.1 | - | 4 | 1326 | 143 | ||
| AK398239.1 | - | 4 | 1326 | 143 | ||
| AK398248.1 | - | 4 | 1326 | 143 | ||
| AK398250.1 | - | 4 | 1326 | 143 | ||
| AK398258.1 | - | 4 | 1326 | 143 | ||
| AK398284.1 | - | 4 | 1326 | 143 | ||
| AK398498.1 | - | 4 | 1326 | 143 | ||
| AK399373.1 | - | 4 | 1326 | 143 | ||
| AK399526.1 | - | 4 | 1329 | 143 | ||
| AK399541.1 | - | 4 | 1327 | 143 | ||
| AK400767.1 | - | 4 | 1326 | 143 | ||
| AK400851.1 | - | 4 | 1326 | 143 | ||
| AK400955.1 | - | 4 | 1326 | 143 | ||
| AK400957.1 | - | 4 | 1326 | 143 | ||
| AK400965.1 | - | 4 | 1326 | 143 | ||
| AK400976.1 | - | 4 | 1339 | 143 | ||
| AK400990.1 | - | 4 | 1326 | 143 | ||
| AK400992.1 | - | 4 | 1327 | 143 | ||
| AK400994.1 | - | 4 | 1326 | 143 | ||
| AK400996.1 | - | 4 | 1326 | 143 | ||
| AK401002.1 | - | 4 | 1326 | 143 | ||
| AK401005.1 | - | 4 | 1326 | 143 | ||
| AK401006.1 | - | 4 | 1330 | 143 | ||
| AK401009.1 | - | 4 | 1325 | 143 | ||
| AK401018.1 | - | 4 | 1326 | 143 | ||
| AK401024.1 | - | 4 | 1326 | 143 | ||
| DQ225365.1 | - | 4 | 1349 | 143 | ||
| Gene Symbol: LOC100127131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355375.4 | XP_003355423.1 | 1428 | 143 | ||
| XM_013988248.2 | XP_013843702.1 | 1532 | 143 | |||
| XM_021091599.1 | XP_020947258.1 | 1466 | 143 | |||
| GenBank mRNA | AK233315.1 | - | 4 | 1386 | 143 | |
| AK234719.1 | - | 3 | 1159 | 143 | ||
| AK235091.1 | - | 4 | 1330 | 143 | ||
| AK235194.1 | - | 4 | 1330 | 143 | ||
| AK235808.1 | - | 4 | 1330 | 143 | ||
| AK236027.1 | - | 4 | 1330 | 143 | ||
| AK236304.1 | - | 4 | 1330 | 143 | ||
| AK392964.1 | - | 4 | 1392 | 143 | ||
| AK394737.1 | - | 6 | 1848 | 143 | ||
| AK396512.1 | - | 4 | 1365 | 143 | ||
| AK396527.1 | - | 4 | 1365 | 143 | ||
| AK398145.1 | - | 4 | 143 | |||
| AK399556.1 | - | 4 | 1377 | 143 | ||
| Gene Symbol: TUBA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315710.1 | NP_001302639.1 | 4 | 1361 | 143 | |
| XM_005655549.2 | XP_005655606.1 | 1573 | 143 | |||
| GenBank mRNA | AK230749.1 | - | 3 | 1561 | 143 | |
| AK234105.1 | - | 4 | 1327 | 143 | ||
| AK236136.1 | - | 4 | 1323 | 143 | ||
| AK237930.1 | - | 4 | 1513 | 143 | ||
| AK350549.1 | - | 4 | 1444 | 143 | ||
| AK389611.1 | - | 4 | 1418 | 143 | ||
| AK391677.1 | - | 4 | 1325 | 143 | ||
| AK392448.1 | - | 4 | 1325 | 143 | ||
| AK399478.1 | - | 4 | 1324 | 143 | ||
| AK400102.1 | - | 4 | 1329 | 143 | ||
| DQ084489.1 | - | 4 | 1361 | 143 | ||
More Information
Set Membership: |
|

TUBA1B
Genomic Map