Assay Details
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134352.2 | NP_001127824.1 | 3 - 4 | 883 | 110 | |
XM_021100283.1 | XP_020955942.1 | 1126 | 110 | |||
XM_021100284.1 | XP_020955943.1 | 1399 | 110 | |||
GenBank mRNA | AK343588.1 | - | 3 - 4 | 1000 | 110 | |
EU669073.2 | - | 3 - 4 | 883 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
