Assay Details
| Gene Symbol: SLC26A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135966.1 | NP_001129438.1 | 10 - 11 | 1145 | 57 | |
| GenBank mRNA | AK233056.1 | - | 5 - 6 | 441 | 57 | |
| AK351373.1 | - | 3 - 4 | 200 | 57 | ||
| AK393373.1 | - | 6 - 7 | 586 | 57 | ||
| AK399849.1 | - | 10 - 11 | 1160 | 57 | ||
| FJ217220.1 | - | 10 - 11 | 1145 | 57 | ||
More Information
Set Membership: |
|

SLC26A11
Genomic Map