Assay Details
| Gene Symbol: PDZK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001769.1 | NP_001001769.1 | 2 | 254 | 102 | |
| XM_005656118.3 | XP_005656175.1 | 378 | 102 | |||
| XM_021093593.1 | XP_020949252.1 | 379 | 102 | |||
| GenBank mRNA | AY626348.1 | - | 2 | 254 | 102 | |
More Information
Set Membership: |
|

PDZK1IP1
Genomic Map